Learn More
Corning™ PETG Baffled Erlenmeyer Flask with Vent Cap
BIOPHARMAGREENER_CHOICE
Constructed of optically clear polyethylene terephthalate (PETG) and ideal for shaker culture applications.
Brand:  Corning™ 431285
 View more versions of this product
Product Code. 15803885
  • 544.00 € / Pack of 4
Estimated Shipment: 26-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
The Corning™ Erlenmeyer flask is designed to help scale-up suspension culture with space efficiency in mind. The flasks are ready-to-use, ideal for shaker culture applications, and storage.
  • Available with baffled bottom
  • Molded-in graduations for accuracy
  • Large vent cap for continuous gas exchange while preventing leakage
  • Highest Sterility Assurance Level (SAL) of 10−6
  • Nonpyrogenic and RNase-/DNase-free
  • Made from optically-clear polyethylene terephthalate(PTEG)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Nonpyrogenic, DNase/RNase-free
Polypropylene
4/Cs.
Fernbach
Vented
Orange
5 L
1/Pk.
Sterile
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Disclaimer: This product is not intended to mitigate the presence of microorganisms on surfaces or in the environment, where such organisms can be deleterious to humans or the environment.