missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Storage solution for pH electrodes
- 500mL bottle
Specifications
Specifications
pH | 2 |
Quantity | 500 mL |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
pH Buffer Solution, Jenway™ >
Spot an opportunity for improvement?Share a Content Correction