Learn More
pH Buffer Solution pH 5, Fisher Chemical™
59.45 € - 229.00 €
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
12850317
|
Reagecon
105025MF |
1 L |
59.45 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
12819826
|
Reagecon
BPH39 |
10 L |
229.00 €
10 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Mercury free buffers at 25°C (pH4 and pH10 are colored) with extended shelf life. Supplied with a detailed certificate of analysis, which outlines traceability to NIST. Temperature dependence data is printed on label as are lot numbers and expiry dates.Specifications
Undesignated | |
5 |