missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ pH Electrode with BNC Connector

With a pH range of 0 to 13, this pH electrode with BNC connector, is ideal for measuring pH in most laboratory and scientific environments.

Brand:  Fisherbrand™ RR5573710-003B

 View more versions of this product

Product Code. 11709798

  • 178.00 € / Each
Estimated Shipment: 18-12-2024
to see stock.
Explore more special offers
Add to basket

Description

Description

  • Electrode pH diameter 6 or 8 mm for measurement in small volumes
  • Electrolyte KCl 4M + AgCl
Specifications

Specifications

BNC
pH Electrode
115 mm
-5°C to +100°C
6 mm
Charged or viscous solutions
0 to 13
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers