missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ pH Electrode with BNC Connector

With a pH range of 0 to 13, this pH electrode with BNC connector, is ideal for measuring pH in most laboratory and scientific environments.

114.00 € - 178.00 €

Specifications

Connector Types BNC
Type pH Electrode
View More Specs

Products 3
Product Code Brand Diameter (Metric) Length (Metric) Price Quantity & Availability  
Product Code Brand Diameter (Metric) Length (Metric) Price Quantity & Availability  
11779788
View Documents
Fisherbrand™
RR5733710-003B
12 mm 120 mm
168.00 €
Each
Estimated Shipment: 19-02-2025
to see stock.
Add to basket
 
11759798
View Documents
Fisherbrand™
5673507-003B
8 mm 115 mm
114.00 €
Each
In Stock
to see stock.
Edge
Add to basket
 
11709798
View Documents
Fisherbrand™
RR5573710-003B
6 mm 115 mm
178.00 €
Each
Estimated Shipment: 19-02-2025
to see stock.
Add to basket
 
Description

Description

  • Electrode pH diameter 6 or 8 mm for measurement in small volumes
  • Electrolyte KCl 4M + AgCl
Specifications

Specifications

BNC
pH Electrode
Videos
SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.