Learn More
Fisherbrand™ pH Indicator Paper
Measure pH simply and easily. Fisherbrand™ pH Indicator Papers produce a distinct color which is then matched with the color scale at specified intervals for easy, reliable readings. These ready-to-use indicator paper rolls are manufactured from high quality filter paper in a variety of pH ranges.
22.45 € - 25.21 €
Specifications
Certifications/Compliance | ISO 9001 |
---|---|
Color Indicator | Color Scale Wheel |
Format | Roll |
For Use With (Application) | pH measurement |
Includes | 1 plastic dispensing wheel, 1 color scale, 4 rolls of indicator paper |
Product Code | Brand | pH Range | Graduations | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | pH Range | Graduations | Price | Quantity & Availability | |||||
15384174
|
Fisherbrand™
90207.204231 |
4 to 7 | 0.3 pH | N/A | ||||||
15374174
|
Fisherbrand™
90205.204231 |
0.5 to 5.5 | 0.5 pH |
25.21 €
4 units |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15314184
|
Fisherbrand™
90213.204231 |
9 to 13 | 0.5 pH | N/A | ||||||
15304184
|
Fisherbrand™
90212.204231 |
8 to 10 | 0.2 / 0.3 / 0.4 pH |
22.45 €
4 units |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
pH indicator papers are standard in many applications for easy, rapid and accurage pH readings. The paper rolls are supplied in plastic dispensing reels for easy use: just Tear, Dip & Read. (For weak or buffered solutions, place the strip against the side of a glass test tube, pour in the test solution and read the result through the glass.)
- Supplied in plastic reels to ensure long term stability and protect against environmental conditions
- Color scales perfectly match the reaction color of the paper for easy, accurate readings
- Indicator paper is always ready to use and can be cut into any length
- ISO 9001 quality control scheme ensures optimal quality and reliability
Specifications
ISO 9001 | |
Roll | |
1 plastic dispensing wheel, 1 color scale, 4 rolls of indicator paper | |
pH Indicator Paper | |
5 m | |
7 mm |
Color Scale Wheel | |
pH measurement | |
4 Each | |
16.4 ft. | |
0.28 in. |
Safety and Handling
missing translation for 'shelfLife' : 3 Years
Your input is important to us. Please complete this form to provide feedback related to the content on this product.