Learn More
Fisherbrand™ pH Indicator Paper
Measure pH simply and easily. Fisherbrand™ pH Indicator Papers produce a distinct color which is then matched with the color scale at specified intervals for easy, reliable readings. These ready-to-use indicator paper rolls are manufactured from high quality filter paper in a variety of pH ranges.
Brand: Fisherbrand™ 90212.204231
Description
pH indicator papers are standard in many applications for easy, rapid and accurage pH readings. The paper rolls are supplied in plastic dispensing reels for easy use: just Tear, Dip & Read. (For weak or buffered solutions, place the strip against the side of a glass test tube, pour in the test solution and read the result through the glass.)
- Supplied in plastic reels to ensure long term stability and protect against environmental conditions
- Color scales perfectly match the reaction color of the paper for easy, accurate readings
- Indicator paper is always ready to use and can be cut into any length
- ISO 9001 quality control scheme ensures optimal quality and reliability
Specifications
ISO 9001 | |
8 to 10 | |
pH measurement | |
1 plastic dispensing wheel, 1 color scale, 4 rolls of indicator paper | |
pH Indicator Paper | |
5 m | |
7 mm |
Color Scale Wheel | |
Roll | |
0.2 / 0.3 / 0.4 pH | |
4 Each | |
16.4 ft. | |
0.28 in. |
Product Suggestions
Customers who viewed this item also viewed
Your input is important to us. Please complete this form to provide feedback related to the content on this product.