missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ pH Indicator Paper Rolls
Provides rapid and precise pH readings. Fisherbrand™ pH Indicator Paper Rolls includesa color scale printed on the cover for convenience.
13.72 € - 71.91 €
Product Code | Brand | pH Range | Format | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | pH Range | Format | Price | Quantity & Availability | ||||||
15565669
|
Fisherbrand™
PH010R |
0 to 10 | Roll | N/A | N/A | N/A | |||||
15595669
|
Fisherbrand™
PH111R/4 |
0 to 11 | Roll | N/A | N/A | N/A | |||||
15535679
|
Fisherbrand™
PH114Z |
1 to 14 | Refill | N/A | N/A | N/A | |||||
15545679
|
Fisherbrand™
PH590R |
5.5 to 9 | Roll | N/A | N/A | N/A | |||||
15555679
|
Fisherbrand™
PH680R/4 |
6.4 to 8 | Roll |
71.91 €
Pack of 4 |
In Stock
Log in to see stock. |
||||||
Description
- Filter paper strips soaked with one or several coloured indicators of pH
- Economical selection of indicator papers cover the entire range of pH 0 to 14
- Available in a variety of formats to meet individual needs
- Reference chart shows colour changes in steps of pH2
Specifications
Color scale printed on cover | |
5 m |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisherbrand™ pH Indicator Paper Rolls