Learn More
Cowie Technology™ PTFE Joint Adapter
Cowie Technology™ PTFE Joint Adapters are grease-free and create high vacuum seals for standard taper ground glass joints.
Brand:  Cowie Technology™ 021.2434
 View more versions of this product
Product Code. 10465252
  • 21.75 € / Each
Estimated Shipment: 20-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Grease-free
  • High vacuum seals for standard taper ground glass joints
  • Substantial shoulder for ease of handling and repeated use
  • High vacuum performance (leakage rate is less than 1 x 10-4 toor, litre, sec.-1) is attained using transverse sealing rings.
  • Use up to 280°C
Specifications
Specifications
1 Each
34/35
NS 24/29, NS 34/35
PTFE Joint Adapters
24/29