missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cowie Technology™ PTFE Joint Adapter
Cowie Technology™ PTFE Joint Adapters are grease-free and create high vacuum seals for standard taper ground glass joints.
11.45 € - 21.75 €
Product Code | Brand | Standard Taper Joint No | Cone Size | Joint Size | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Standard Taper Joint No | Cone Size | Joint Size | Price | Quantity & Availability | |||
10211762
|
Cowie Technology™
021.1019 |
NS 10/19, NS 19/26 | 19/26 | 10/19 |
11.45 €
Each |
Estimated Shipment: 31-03-2025
Log in to see stock. |
|||
10363452
|
Cowie Technology™
021.1419 |
NS 14/23, NS 19/26 | 19/26 | 14/23 |
11.65 €
Each |
Estimated Shipment: 30-01-2025
Log in to see stock. |
|||
10271852
|
Cowie Technology™
021.1934 |
NS 19/26, NS 34/35 | 34/35 | 19/26 |
21.75 €
Each |
Estimated Shipment: 20-02-2025
Log in to see stock. |
|||
10313602
|
Cowie Technology™
021.1929 |
NS 19/26, NS 29/32 | 29/32 | 19/26 |
18.25 €
Each |
Estimated Shipment: 30-01-2025
Log in to see stock. |
|||
10363692
|
Cowie Technology™
021.2429 |
NS 24/29, NS 29/32 | 29/32 | 24/29 |
21.60 €
Each |
Estimated Shipment: 30-01-2025
Log in to see stock. |
|||
10150382
|
Cowie Technology™
021.2934 |
NS 29/32, NS 34/35 | 34/35 | 29/32 |
21.75 €
Each |
Estimated Shipment: 20-01-2025
Log in to see stock. |
|||
10465252
|
Cowie Technology™
021.2434 |
NS 24/29, NS 34/35 | 34/35 | 24/29 |
21.75 €
Each |
Estimated Shipment: 20-01-2025
Log in to see stock. |
|||
Description
- Grease-free
- High vacuum seals for standard taper ground glass joints
- Substantial shoulder for ease of handling and repeated use
- High vacuum performance (leakage rate is less than 1 x 10-4 toor, litre, sec.-1) is attained using transverse sealing rings.
- Use up to 280°C
Specifications
1 Each | |
PTFE Joint Adapters |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title