missing translation for 'onlineSavingsMsg'
Learn More

Christeyns™ Phago'Derm Pure Soap

Phago'Derm Pure is intended for the simple washing of hands and forearms in intensive use.

Brand:  Christeyns™ C65074


 View more versions of this product

Product Code. 17860245

  • 84.85 € / 12 L
Estimated Shipment: 31-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Also suitable for patient washing and preoperative showering
  • Recommended for sensitive and reactive skin
  • Square bottle with mounted pump
Specifications

Specifications

Hand Sanitizer Gel
Liquid
1 L
Colorless
12 Airless Bottles
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • PHAGO'SANIT

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1

Hazard Statement
  • H318-Causes serious eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

Supplemental information
  • MIXTURE LIST-Contains : Lactic acid, aceticacid, hydrogen peroxide, Alkyl polyglycoside

SDS
Documents

Documents

Certificates
Special Offers

Special Offers