missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Christeyns™ Phago'Derm Pure Soap
Phago'Derm Pure is intended for the simple washing of hands and forearms in intensive use.
74.15 € - 84.85 €
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
16873083
|
Christeyns™
65072 |
500 mL |
74.15 €
Pack of 20 |
Estimated Shipment: 02-01-2025
Log in to see stock. |
|||||
17860245
|
Christeyns™
|
1 L |
84.85 €
12 L |
Estimated Shipment: 30-12-2024
Log in to see stock. |
|||||
Description
- Also suitable for patient washing and preoperative showering
- Recommended for sensitive and reactive skin
- Square bottle with mounted pump
Specifications
Colorless | |
Liquid |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- X20 Phago'Derm Pure mild soap 500ml
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : Ethanol, 1-Propanaminium, Amines, C12-14