missing translation for 'onlineSavingsMsg'
Learn More

Christeyns™ Phago'Surf 2D

Phago'Surf 2D is a concentrated liquid intended for the cleaning and disinfection of floors, surfaces and materials in high-risk environments.

Brand:  Christeyns™ C60423


 View more versions of this product

Product Code. 17860265

  • 115.00 € / 10 L
Estimated Shipment: 03-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Effective cleaner and broad-spectrum disinfectant (bactericidal, yeasticidal, fungicidal, tuberculocidal and active on viruses)
  • Requires rinsing on surfaces likely to come into direct contact with foodstuffs (in accordance with the decree of September 8, 1999)
  • Biocidal product registered in TP2 TP3 and TP4
  • Fast drying, streak-free
  • No scent
  • 'Food contact' product
Specifications

Specifications

Disinfectant Detergent
Concentrated Liquid
Unscented
2 Cans
No
Neutral
Non-sterile
5 L
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • PHAGOSURF 2D

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • ACUTE AQUATIC Acute 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H400-Very toxic to aquatic life.
  • H411-Toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

Supplemental information
  • MIXTURE LIST-Contains : N-(3-Aminopropyl)-N-dodecylpropane-1,3-diamine, Didecyldimethylammonium chloride, Ethoxylated Laury

SDS
Documents

Documents

Certificates
Special Offers

Special Offers