missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Phase Separating Paper, Grade: 902

Used for the separation of aqueous solutions from organic solutions

Brand:  Fisherbrand™ 0902A00004

51.99 EUR valid until 2025-07-13
Use promo code "24135" to get your promotional price.


 View more versions of this product

Product Code. 11794033

  • 62.60 € / Pack of 100
Estimated Shipment: 19-05-2025
to see stock.
Explore more special offers
Add to basket

Description

Description

  • Hydrophobic, with silicone impregnated paper
  • Organic phase permeates filter whereas aqueous phase, solids components are retained
  • Usable as disposable separating funnel
  • Density of organic phase has no influence on separating process
Specifications

Specifications

85 g/m2
Silicone
Hydrophobic
Phase Separating Paper
DIN 53137
Separating aqueous solutions from organic solutions, Usable as disposable separating funnel
0.17 mm
150 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ Phase Separating Paper, Grade: 902 > Dia.: 150mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.