missing translation for 'onlineSavingsMsg'
Learn More

Phenol Red Indicator, ACS Reagent, Honeywell Fluka™

Indicator, ACS reagent

66.39 € - 346.66 €

Chemical Identifiers

CAS 143-74-8
Molecular Formula C19H14O5S
Molecular Weight (g/mol) 354.38
MDL Number MFCD00003552
InChI Key BELBBZDIHDAJOR-UHFFFAOYSA-N
Synonym Phenolsulfonphthalein
PubChem CID 4766
ChEBI CHEBI:31991
IUPAC Name 4-[3-(4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol
SMILES OC1=CC=C(C=C1)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15653830
View Documents
Honeywell-Fluka
32661-10G
10 g Glass Bottle
66.39 €
10g
Estimated Shipment: 14-02-2025
to see stock.
Add to basket
 
15663830
View Documents
Honeywell-Fluka
32661-25G
25 g Glass Bottle
92.44 €
25g
Estimated Shipment: 28-11-2024
to see stock.
Add to basket
 
15643830
View Documents
Honeywell-Fluka
32661-100G
100 g Glass Bottle
346.66 €
100g
Estimated Shipment: 02-01-2025
to see stock.
Add to basket
 
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

143-74-8
354.38
BELBBZDIHDAJOR-UHFFFAOYSA-N
4766
4-[3-(4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol
C19H14O5S
MFCD00003552
Phenolsulfonphthalein
CHEBI:31991
OC1=CC=C(C=C1)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1

Specifications

143-74-8
MFCD00003552
326470
BELBBZDIHDAJOR-UHFFFAOYSA-N
4-[3-(4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol
4766
354.38g/mol
6.8 to 8.2 (yellow to red)
Powder
C19H14O5S
NONH for all modes of transport
Phenolsulfonphthalein
OC1=CC=C(C=C1)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1
354.38
CHEBI:31991
ACS Reagent
Red
Phenol Red
Videos
Safety and Handling

Safety and Handling

P261-P305 + P351 + P338

H315-H319-H335

missing translation for 'einecsNumber' : 205-609-7

missing translation for 'rtecsNumber' : SJ7490000

SDS
Documents

Documents

Product Certifications