missing translation for 'onlineSavingsMsg'
Learn More

Phenol Red, Pure, Indicator Grade, Fisher Chemical™

39.05 € - 367.00 €

Chemical Identifiers

CAS 143-74-8
Molecular Formula C19H14O5S
Molecular Weight (g/mol) 354.38
MDL Number MFCD00003552
InChI Key BELBBZDIHDAJOR-UHFFFAOYSA-N
PubChem CID 4766
ChEBI CHEBI:31991
SMILES OC1=CC=C(C=C1)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10315860
View Documents
Fisher Chemical
P/2420/44
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10111580
View Documents
Fisher Chemical
P/2420/46
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11463723
View Documents
Fisher Chemical
P/2420/53
500 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications

Chemical Identifiers

143-74-8
354.38
BELBBZDIHDAJOR-UHFFFAOYSA-N
CHEBI:31991
C19H14O5S
MFCD00003552
4766
OC1=CC=C(C=C1)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1

Specifications

300°C
C19H14O5S
BELBBZDIHDAJOR-UHFFFAOYSA-N
3,3-bis(4-hydroxyphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione
4766
354.38g/mol
Brown/Red
143-74-8
MFCD00003552
OC1=CC=C(C=C1)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1
354.38
CHEBI:31991
Glass Jar
Powder
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Phenol Red

Signal Word
  • Warning

Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.