Learn More
Phenol, Saturated (pH 4.3, Liq.), Fisher BioReagents
73.52 € - 96.26 €
Chemical Identifiers
CAS | 108-95-2 |
---|---|
Molecular Formula | C6H6O |
Molecular Weight (g/mol) | 94.11 |
MDL Number | MFCD00002143 |
InChI Key | ISWSIDIOOBJBQZ-UHFFFAOYSA-N |
Synonym | carbolic acid, hydroxybenzene, phenic acid, phenylic acid, oxybenzene, benzenol, phenyl hydrate, monophenol, phenyl hydroxide, phenylic alcoholShow More |
PubChem CID | 996 |
ChEBI | CHEBI:15882 |
IUPAC Name | phenol |
SMILES | OC1=CC=CC=C1 |
Description
Phenol saturated with Tris buffer is used for purifying RNA.Chemical Identifiers
108-95-2 | |
94.11 | |
ISWSIDIOOBJBQZ-UHFFFAOYSA-N | |
996 | |
phenol |
C6H6O | |
MFCD00002143 | |
carbolic acid, hydroxybenzene, phenic acid, phenylic acid, oxybenzene, benzenol, phenyl hydrate, monophenol, phenyl hydroxide, phenylic alcoholShow More | |
CHEBI:15882 | |
OC1=CC=CC=C1 |
Specifications
108-95-2 | |
C6H6O | |
1.05g/cm³ | |
ISWSIDIOOBJBQZ-UHFFFAOYSA-N | |
phenol | |
996 | |
≥99.0% | |
4.3 | |
Phenol, Saturated |
≥99.0 | |
MFCD00002143 | |
carbolic acid, hydroxybenzene, phenic acid, phenylic acid, oxybenzene, benzenol, phenyl hydrate, monophenol, phenyl hydroxide, phenylic alcohol | |
OC1=CC=CC=C1 | |
94.11 | |
CHEBI:15882 | |
Pass Test | |
Liquid |
Safety and Handling

DANGER!
Emergency Overview
Combustible liquid. Causes burns by all exposure routes. May cause central nervous system effects. Possible risks of irreversible effects. Danger of serious damage to health by prolonged exposure. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Immediate medical attention is required. Move to fresh air.
NFPA
Health:4
Flammability:2
Instability:0
missing translation for 'dotInformation' : DOT Class 6.1, : Poison
Recommended Storage : 4°C
- Phenol, Saturated (pH 4.3)
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
- Germ cell mutagenicity Category 2
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H314-Causes severe skin burns and eye damage.
- H373-May cause damage to organs.
- H341-Suspected of causing genetic defects.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
Supplemental information
- MIXTURE LIST-Contains : Phenol, 1,2-Benzenedicarboxylic acid, monopotassium salt
Your input is important to us. Please complete this form to provide feedback related to the content on this product.