missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ phi29 DNA Polymerase (10 U/μL)

Thermo Scientific phi29 DNA Polymerase is a highly processive polymerase featuring strong strand displacement activity, which allows for highly efficient isothermal DNA amplification.

Brand:  Thermo Scientific™ EP0092

 View more versions of this product

Product Code. 10242410

  • 319.00 € / 1000 units

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

Stand-alone enzyme
phi29 DNA Polymerase
• phi29 DNA Polymerase (1,000 units at 10 U/μL)
• 10X Reaction Buffer for phi29 DNA Polymerase
• Store at –15 to –25°C.
Dry Ice
4 hours
MDA-WGA, RCA
1,000 units
30°C
10 U/μL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2807181 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberEP0092
Lot Number2851758 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog NumberEP0092
Lot Number2851785 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog NumberEP0092
Lot Number2901634 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog NumberEP0092
Lot Number2901650 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog NumberEP0092
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ phi29 DNA Polymerase (10 U/μL) > 1000 U

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.