missing translation for 'onlineSavingsMsg'
Learn More
Phloroglucinol, 99+%, anhydrous, Thermo Scientific Chemicals Product Code.: 10002283

Phloroglucinol, 99+%, anhydrous, Thermo Scientific Chemicals

Product Code. 10002283
25 g, Glass bottle
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10002283

Brand: Thermo Scientific Acros 241760250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-73-6
Molecular Formula C6H6O3
Molecular Weight (g/mol) 126.1
InChI Key QCDYQQDYXPDABM-UHFFFAOYSA-N
Synonym phloroglucinol, 1,3,5-benzenetriol, 1,3,5-trihydroxybenzene, phloroglucin, phloroglucine, spasfon-lyoc, s-trihydroxybenzene, benzene-s-triol, 5-hydroxyresorcinol, benzene, trihydroxyShow MoreShow Less
PubChem CID 359
ChEBI CHEBI:16204
IUPAC Name benzene-1,3,5-triol
SMILES C1=C(C=C(C=C1O)O)O

Specifications

Melting Point 215.0°C to 218.0°C
Color White to Beige
Flash Point 100°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (HPLC)
Packaging Glass bottle
Linear Formula (OH)3C6H3
Quantity 25 g
Fieser 01,47; 06,147
Merck Index 15,7439
Solubility Information Solubility in water: 10g/L (20°C). Other solubilities: soluble in acetone ethanol, ether and pyridine
Formula Weight 126.1
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material Phloroglucinol
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0463323 Certificate TypeCertificate of Analysis Date14/06/2024 Catalog Number241765000,241761000,241760250
Lot NumberA0450555 Certificate TypeCertificate of Analysis Date21/02/2024 Catalog Number241765000,241761000,241760250
Lot NumberA0449598 Certificate TypeCertificate of Analysis Date26/01/2023 Catalog Number241765000,241761000,241760250
Lot NumberA0392002 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number241765000,241761000,241760250
Lot NumberA0406388 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number241765000,241761000,241760250
compliance-icons
Product Identifier
  • Phloroglucinol, anhydrous
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Phloroglucinol, 99+%, anhydrous, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.