Learn More
Phloroglucinol dihydrate, 98%, Thermo Scientific Chemicals
Description
Phloroglucinol dihydrate is utilized in the preparation of active pharmaceutical ingredients such as flopropione. It acts as a reagent for the Tollens’ test for pentoses. It is mixed with hydrochloric acid and plays an important role for the detection of lignin (Wiesner test). It is involved in the analysis and qualitative detection of free hydrochloric acid in gastric juice by using Gunzburg reagent. Further, it serves as a coupling agent in printing. It is also employed as a reagent for pentoses, pentosans, aldehydes and as a decalcifier of bone specimens in microscopy.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phloroglucinol dihydrate is utilized in the preparation of active pharmaceutical ingredients such as flopropione. It acts as a reagent for the Tollens’ test for pentoses. It is mixed with hydrochloric acid and plays an important role for the detection of lignin (Wiesner test). It is involved in the analysis and qualitative detection of free hydrochloric acid in gastric juice by using Gunzburg reagent. Further, it serves as a coupling agent in printing. It is also employed as a reagent for pentoses, pentosans, aldehydes and as a decalcifier of bone specimens in microscopy.
Solubility
Slightly soluble in water.
Notes
Light sensitive. Incompatible with acid chlorides, acid anhydrides, bases and oxidizing agents.

Specifications
Specifications
Melting Point | 214°C to 217°C |
Odor | Odorless |
Quantity | 25 g |
Sensitivity | Light sensitive |
Merck Index | 14,7328 |
Solubility Information | Slightly soluble in water. |
Formula Weight | 162.14 (126.11 Anhydrous) |
Percent Purity | 98% |
Chemical Name or Material | Phloroglucinol dihydrate |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
10252348 | Certificate of Analysis | 10/09/2024 | A13034.14,A13034.36,A13034.22 |
10244547 | Certificate of Analysis | 16/11/2022 | A13034.14,A13034.36,A13034.22 |
10123978 | Certificate of Analysis | 22/11/2021 | A13034.14,A13034.36,A13034.22 |
10194586 | Certificate of Analysis | 22/11/2021 | A13034.14,A13034.36,A13034.22 |
10103263 | Certificate of Analysis | 22/11/2021 | A13034.14,A13034.36,A13034.22 |
Safety and Handling
- Phloroglucinol dihydrate
- LONG-TERM AQUATIC HAZARD Chronic 3
- H412-Harmful to aquatic life with long lasting effects.
- P273-Avoid release to the environment.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : Phloroglucinol dihydrate
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.