Learn More
Phospho-MEK1 (Thr386) Rabbit, anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 28935-1-AP-20UL
This item is not returnable.
View return policy
Description
MEK1 is also named as MAP2K1 (mitogen-activated protein kinase kinase 1), MAPKK1, PRKMK1, MKK1 and belongs to the MAP kinase kinase subfamily. It is a dual-specificity kinase that mediate ERK1 and ERK2 activation during adhesion and growth factor signaling(PMID:19219045). It also plays an essential role in extra-embryonic ectoderm during placentogenesis. MEK1 has 2 isoforms produced by alternative splicing with the molecular weight of 43 kDa and 41 kDa. MEK1 may form a heterodimer with Mek2.(PMID:21144847,19219045).
Specifications
Phospho-MEK1 (Thr386) | |
Polyclonal | |
Unconjugated | |
20 μL | |
5604 | |
Liquid |
Western Blot, ELISA | |
200 μg/mL | |
Rabbit | |
Primary | |
Human |