missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-MEK1 (Thr386) Rabbit, anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
130.00 € - 350.00 €
Specifications
Antigen | Phospho-MEK1 (Thr386) |
---|---|
Concentration | 200 μg/mL |
Applications | Western Blot, ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17211426
|
Proteintech
28935-1-AP-20UL |
20 μL |
130.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17201426
|
Proteintech
28935-1-AP-100UL |
100 μL |
350.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MEK1 is also named as MAP2K1 (mitogen-activated protein kinase kinase 1), MAPKK1, PRKMK1, MKK1 and belongs to the MAP kinase kinase subfamily. It is a dual-specificity kinase that mediate ERK1 and ERK2 activation during adhesion and growth factor signaling(PMID:19219045). It also plays an essential role in extra-embryonic ectoderm during placentogenesis. MEK1 has 2 isoforms produced by alternative splicing with the molecular weight of 43 kDa and 41 kDa. MEK1 may form a heterodimer with Mek2.(PMID:21144847,19219045).
Specifications
Phospho-MEK1 (Thr386) | |
Western Blot, ELISA | |
Unconjugated | |
Rabbit | |
5604 |
200 μg/mL | |
Polyclonal | |
Liquid | |
Human | |
Primary |