Learn More
Phosphomolybdic acid, ammonium salt hydrate, Thermo Scientific Chemicals
72.16 € - 745.71 €
Chemical Identifiers
CAS | 54723-94-3 |
---|---|
Molecular Formula | H80Mo12N3O41P-6 |
Molecular Weight (g/mol) | 1960.994 |
MDL Number | MFCD00150423 |
InChI Key | BIQVMZMYTFVCJZ-UHFFFAOYSA-H |
Synonym | ammonium phosphomolybdate hydrate |
PubChem CID | 131664298 |
IUPAC Name | azane;molybdenum;trihydroxide;phosphate;tetratriacontahydrate |
SMILES | N.N.N.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.[OH-].[OH-].[OH-].[O-]P(=O)([O-])[O-].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10420161
|
Thermo Scientific Acros
317100500 |
50 g | Plastic bottle |
72.16 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10137763
|
Thermo Scientific Acros
317102500 |
250 g | Plastic bottle |
193.97 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10678824
|
Thermo Scientific Acros
317100010 |
1 kg | Plastic bottle |
745.71 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagentChemical Identifiers
54723-94-3 | |
1960.994 | |
BIQVMZMYTFVCJZ-UHFFFAOYSA-H | |
131664298 | |
N.N.N.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.[OH-].[OH-].[OH-].[O-]P(=O)([O-])[O-].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo] |
H80Mo12N3O41P-6 | |
MFCD00150423 | |
ammonium phosphomolybdate hydrate | |
azane;molybdenum;trihydroxide;phosphate;tetratriacontahydrate |
Specifications
Yellow | |
H80Mo12N3O41P-6 | |
Powder | |
ammonium phosphomolybdate hydrate | |
BIQVMZMYTFVCJZ-UHFFFAOYSA-H | |
azane;molybdenum;trihydroxide;phosphate;tetratriacontahydrate | |
131664298 | |
Phosphomolybdic acid, ammonium salt hydrate |
54723-94-3 | |
(NH4)3PMo12O40·xH2O | |
MFCD00150423 | |
Solubility in water: insoluble. | |
N.N.N.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.[OH-].[OH-].[OH-].[O-]P(=O)([O-])[O-].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo].[Mo] | |
1960.994 | |
1876.35 |
Safety and Handling
GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.