missing translation for 'onlineSavingsMsg'
Learn More

Phosphonitrilic chloride trimer, 98%, Thermo Scientific Chemicals

19.60 € - 122.61 €

Chemical Identifiers

CAS 940-71-6
Molecular Formula Cl6N3P3
Molecular Weight (g/mol) 347.65
MDL Number MFCD00006474
InChI Key UBIJTWDKTYCPMQ-UHFFFAOYSA-N
Synonym phosphonitrilic chloride trimer, hexachlorocyclotriphosphazene, hexachlorophosphazene, triphosphonitrilic chloride, triphosphonitrile chloride, hexachlorotriphosphonitrile, cyclophosphazene dichloride trimer, hexachlorocyclophosphazatriene, hexachlorocyclotriphosphazatriene, phosphononitrilic chloride trimer
PubChem CID 220225
IUPAC Name 2,2,4,4,6,6-hexachloro-1,3,5-triaza-2$l^{5},4$l^{5},6$l^{5}-triphosphacyclohexa-1,3,5-triene
SMILES N1=P(N=P(N=P1(Cl)Cl)(Cl)Cl)(Cl)Cl
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10367400
View Documents
Thermo Scientific Acros
174280050
5 g Glass bottle
19.60 €
5g
Please to purchase this item. Need a web account? Register with us today!
10572771
View Documents
Thermo Scientific Acros
174280250
25 g Glass bottle
55.07 €
25g
Please to purchase this item. Need a web account? Register with us today!
10530414
View Documents
Thermo Scientific Acros
174281000
100 g Glass bottle
122.61 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Certificates

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Phosphonitrilic chloride trimer, 98%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.