Learn More
Grant Instruments™ DEN-600 - Densitometer, Portable Battery/Mains, Cell Photometer Turbidity 0 - 16 McF & 0.3 Abs
The DEN-600 is a compact, portable, rechargeable battery powered photometer. It’s 600 nm wavelength optical system allows you to measure estimated total number of cells (OD600), turbidity measurement method (McFarland (McF)), protein concentration measurement (Bradford protein assay method)
Brand:  Grant Instruments™ DEN-600
Product Code. 17541136
  • 990.00 € / Each
Explore more special offers
Description
Description
The device serves as an inexpensive alternative to a spectrophotometer, which is commonly used for these applications. As the DEN-600 is battery powered and compact, it can be comfortably located in a biosafety cabinet, anaerobic chamber or quickly relocated.
The cell holder accommodates standard 10 mm path cuvettes, round bottom, conical vials or falcon tubes.
USB connectivity and DEN software allow for data transfer, data processing and calculation.
Applications
  • Cell concentration measurement
  • Cell growth data estimation
  • Log phase estimation for microbial cells induction
  • Competent cell preparation
  • Bradford protein assay method
  • Antibiotic susceptibility testing
  • Inhibitory tests
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Photometer
±0.006 at 1 Abs, ±0.1 at 0 to 8 McF
For 10 mm path cuvettes, round bottom and falcon tubes
65 mm
600 nm
120 mm
LED
145 mm
120/240 V
LED
0.5 kg
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Grant Instruments™ DEN-600 - Densitometer, Portable Battery/Mains, Cell Photometer Turbidity 0 - 16 McF & 0.3 Abs > DEN-600 - Densitometer, portable battery/mains, cell photometer turbidity 0 - 16 McF & 0.3 Abs,wavelength 600nm, for 10mm path cuvettes, round bottom, & falcon tubes