Learn More
Palintest™ Photometer Tablet Reagents
Photometer reagents in convenient tablet form. Palintest™; Photmoeter Tablet Reagents are available for a variety of water testing needs.
Brand: Palintest™ PM162
This item is not returnable.
View return policy
Description
- Convenient tablet form.
- Easy to use.
- Available for a variety of water testing needs.
Specifications
50 | |
0 to 250mg/L | |
Chlorine HR Test Kit |
50/Pk. | |
50 Tests |
Product Suggestions
Customers who viewed this item also viewed
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- TEST CHLORE TOTAL 0-250MG/L
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
Hazard Statement
- H319-Causes serious eye irritation.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H315-Causes skin irritation.
Precautionary Statement
- P264-Wash contaminated skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P330-Rinse mouth.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : adipic acid, citricacid, potassium iodide
Product Content Correction
Product Title
Palintest™ Photometer Tablet Reagents > Range: 0 to 250mg/L
Spot an opportunity for improvement?Share a Content Correction