missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Palintest™ Photometer Tablet Reagents
Photometer reagents in convenient tablet form. Palintest™; Photmoeter Tablet Reagents are available for a variety of water testing needs.
51.85 € - 159.10 €
Product Code | Brand | Type | Range | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Range | Price | Quantity & Availability | |||||
12581457
|
Palintest™
PM064 |
Chlorine Dioxide LR Test Kit | 0 to 2.5mg/L | N/A | ||||||
13108840
|
Palintest™
PM 065 |
Chlorine Dioxide HR Test Kit | 2.5 to 2mg/L | N/A | ||||||
11903627
|
Palintest™
PM179 |
Fluoride Test Kit | 0 to 1.5mg/L |
71.00 €
Pack of 50 |
Estimated Shipment: 22-04-2025
Log in to see stock. |
|||||
13151451
|
Palintest™
PM 262 |
Organophosphonate (OP) Test Kit | 0 to 20mg/L | N/A | ||||||
11923657
|
Palintest™
PM188 |
Alkalinity Total (Alkaphot) Test Kit | 0 to 500mg/L |
59.70 €
Pack of 50 |
Estimated Shipment: 02-04-2025
Log in to see stock. |
|||||
11993607
|
Palintest™
PM162 |
Chlorine HR Test Kit | 0 to 250mg/L |
51.85 €
Pack of 50 |
Estimated Shipment: 09-04-2025
Log in to see stock. |
|||||
11933627
|
Palintest™
PM175 |
Molybdate HR Test Kit | 0 to 100mg/L | N/A | ||||||
11913657
|
Palintest™
PM168 |
Sulphide Test Kit | 0 to 0.5mg/L |
61.00 €
Pack of 50 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Convenient tablet form.
- Easy to use.
- Available for a variety of water testing needs.
Specifications
50 | |
50/Pk. | |
50 Tests |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Palintest™ Photometer Tablet Reagents