Learn More
Palintest™ Photometer Tablet Reagents
Photometer reagents in convenient tablet form. Palintest™; Photmoeter Tablet Reagents are available for a variety of water testing needs.
Brand:  Palintest™ PM 065
 View more versions of this product
Product Code. 13108840
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Convenient tablet form.
  • Easy to use.
  • Available for a variety of water testing needs.
Specifications
Specifications
50
2.5 to 2mg/L
Chlorine Dioxide HR Test Kit
50/Pk.
50 Tests
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Palintest™ Photometer Tablet Reagents > Range: 2.5 to 2mg/L