missing translation for 'onlineSavingsMsg'
Learn More
Phthalhydrazide, 98%, Thermo Scientific Chemicals Product Code.: 11434030

Phthalhydrazide, 98%, Thermo Scientific Chemicals

Product Code. 11434030
10 g
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy

Product Code. 11434030

Brand: Thermo Scientific Alfa Aesar A14615.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Phthalhydrazide is a reagent used in the synthesis of various pyrazolophthalazine derivatives is and used in medicine double hydrazine phthalocyanine work intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phthalhydrazide is a reagent used in the synthesis of various pyrazolophthalazine derivatives is and used in medicine double hydrazine phthalocyanine work intermediates.

Solubility
Soluble in acetone and acetic acid.

Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1445-69-8
Molecular Formula C8H6N2O2
Molecular Weight (g/mol) 162.15
MDL Number MFCD00006888
InChI Key JNQUJPBFSQETPT-UHFFFAOYNA-N
Synonym 2,3-dihydro-1,4-phthalazinedione, phthalazine-1,4-diol, phtalylhydrazine, 1,2,3,4-tetrahydrophthalazine-1,4-dione, 1,4-phthalazinedione, 2,3-dihydro, 1 2h-phthalazinone, 4-hydroxy, 2,3-dihydro-phthalazine-1,4-dione, phthalhydrazine, phthalylhydrazine, phthaloylhydrazineShow MoreShow Less
PubChem CID 219401
SMILES O=C1N=NC(=O)C2C=CC=CC12

Specifications

Melting Point 342°C to 345°C
Quantity 10 g
Beilstein 135926
IUPAC Name 1,4,4a,8a-tetrahydrophthalazine-1,4-dione
Formula Weight 162.15
Percent Purity 98%
Chemical Name or Material Phthalhydrazide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10251572 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberA14615.18,A14615.09
Lot NumberC4532B Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14615.18,A14615.09
Lot NumberD6241A Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14615.18,A14615.09
Lot Number10207745 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14615.18,A14615.09
Lot Number10221868 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14615.18,A14615.09

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Phthalhydrazide, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.