Learn More
Thermo Scientific™ Phusion Blood Direct PCR Kit
Thermo Scientific Phusion Blood Direct PCR Kit is developed for amplification of DNA from whole blood. It eliminates the need for a separate DNA purification step prior to PCR.
Brand: Thermo Scientific™ F547S
81.25 EUR valid until 2024-12-31
Use promo code "21651" to get your promotional price.
Description
Thermo Scientific Phusion Blood Direct PCR Kit is for use in the amplification of DNA from whole blood. It eliminates the need for a separate DNA purification step prior to PCR. The modified Phusion Hot Start II High-Fidelity DNA Polymerase exhibits high resistance to PCR inhibitors found in blood and retains polymerase activity even with samples containing 40% whole blood.
This DNA polymerase exhibits exceptional accuracy in proofreading and makes the Phusion Blood Direct PCR Kit one of the best choices for demanding applications, such as cloning. The Phusion Blood Direct PCR Kit is versatile and permits analysis of blood samples from various species and with different anticoagulants or storage conditions.
Features
- No need for time-consuming and expensive DNA purification steps
- Simple protocol requires minimal hands-on time
- Extremely short PCR protocol times
- Modified Phusion Hot Start II High-Fidelity DNA Polymerase delivers abundant yields of specific product
Notes
- The optimal annealing temperature for Phusion DNA Polymerases may differ significantly from that of Taq-based polymerases.
Specifications
• Phusion Blood II DNA Polymerase, 40 μL • 2X Phusion Blood PCR Buffer with dNTPs and MgCl2, 1.25 mL • Universal Control Primer Mix, 40 μL • 50 mM EDTA, 0.5 mL • 50 mM MgCl2, 1.5 mL • DMSO, 0.5 mL Store at -20°C. |
|
Direct PCR | |
Fast | |
100 reactions | |
2 U/μL | |
100 Reactions | |
Separate Components | |
DNA |
High | |
Phusion Blood II DNA Polymerase | |
Direct PCR Kit | |
Wet or Dry Ice | |
Built-In Hot Start | |
Blunt | |
Blood |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.