missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Phusion™ HF and GC Buffer Packs

5X Phusion Green buffers include a density reagent and two tracking dyes for direct loading of PCR products on a gel.

Brand:  Thermo Scientific™ F519L

 View more versions of this product

Product Code. 10584578

  • 33.90 € / Pack of 4

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The error rate of Phusion DNA Polymerase in HF Buffer (4.4 x 10-7) is lower than that in GC Buffer (9.5 x 10-7). Therefore, the HF Buffer should be used as the default buffer for high-fidelity amplification. However, GC Buffer can improve the performance of Phusion DNA Polymerase on some difficult or long templates, such as GC-rich templates or those with complex secondary structures. For applications such as microarray or DHPLC, where the DNA templates need to be free of detergents, we recommend using detergent-free reaction buffers.

5X Phusion Green Buffers include a density reagent and two tracking dyes for direct loading of PCR products on a gel. The colored buffers do not interfere with the enzyme performance and are compatible with downstream applications such as DNA sequencing, ligation and restriction digestion.

Two optimized buffers are available for Thermo Scientific Phusion High-Fidelity DNA Polymerases:

  • 5X Phusion HF Buffer (F-518L, F-538L (Phusion Green), F-520L (Detergent-free))
  • 5X Phusion GC Buffer (F-519L, F-539L (Phusion Green), F-521L (Detergent-free))
TRUSTED_SUSTAINABILITY
Specifications

Specifications

• 5X Phusion GC Reaction Buffer (4 x 1.5 mL)
• 50 mM MgCl2 (4 x 1.5 mL)
• DMSO (0.5 mL)

Store at –20°C.
Phusion High-Fidelity DNA Polymerase
4 x 1.5 mL
Standard PCR, High-fidelity PCR
4 x 1.5 mL
Phusion™ GC Buffer Pack
GC Buffer Pack
Wet or Dry Ice
DNA
Colorless
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.