missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Phusion™ HF and GC Buffer Packs

5X Phusion Green buffers include a density reagent and two tracking dyes for direct loading of PCR products on a gel.

33.15 € - 38.26 €

Specifications

Polymerase Phusion High-Fidelity DNA Polymerase
Shipping Condition Wet or Dry Ice
For Use With (Application) Standard PCR, High-fidelity PCR
Starting Material DNA
Volume 4 x 1.5 mL
View More Specs

Products 6
Product Code Brand Product Type Color Price Quantity & Availability  
Product Code Brand Product Type Color Price Quantity & Availability  
10492088
missing translation for 'viewDocuments'
Thermo Scientific™
F518L
HF Buffer Pack Colorless
33.28 €
Pack of 4
Please to purchase this item. Need a web account? Register with us today!
10584578
missing translation for 'viewDocuments'
Thermo Scientific™
F519L
GC Buffer Pack Colorless
33.17 €
Pack of 4
Please to purchase this item. Need a web account? Register with us today!
10226009
missing translation for 'viewDocuments'
Thermo Scientific™
F520L
Detergent-free FF Buffer Pack Colorless
37.97 €
Pack of 4
Please to purchase this item. Need a web account? Register with us today!
10333609
missing translation for 'viewDocuments'
Thermo Scientific™
F521L
Detergent-free GC Buffer Pack Colorless
38.26 €
Pack of 4
Please to purchase this item. Need a web account? Register with us today!
13525170
missing translation for 'viewDocuments'
Thermo Scientific™
F538L
HF Buffer Pack Green
33.28 €
Pack of 4
Please to purchase this item. Need a web account? Register with us today!
13535170
missing translation for 'viewDocuments'
Thermo Scientific™
F539L
GC Buffer Pack Green
33.15 €
Pack of 4
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The error rate of Phusion DNA Polymerase in HF Buffer (4.4 x 10-7) is lower than that in GC Buffer (9.5 x 10-7). Therefore, the HF Buffer should be used as the default buffer for high-fidelity amplification. However, GC Buffer can improve the performance of Phusion DNA Polymerase on some difficult or long templates, such as GC-rich templates or those with complex secondary structures. For applications such as microarray or DHPLC, where the DNA templates need to be free of detergents, we recommend using detergent-free reaction buffers.

5X Phusion Green Buffers include a density reagent and two tracking dyes for direct loading of PCR products on a gel. The colored buffers do not interfere with the enzyme performance and are compatible with downstream applications such as DNA sequencing, ligation and restriction digestion.

Two optimized buffers are available for Thermo Scientific Phusion High-Fidelity DNA Polymerases:

  • 5X Phusion HF Buffer (F-518L, F-538L (Phusion Green), F-520L (Detergent-free))
  • 5X Phusion GC Buffer (F-519L, F-539L (Phusion Green), F-521L (Detergent-free))
Specifications

Specifications

Phusion High-Fidelity DNA Polymerase
Standard PCR, High-fidelity PCR
4 x 1.5 mL
Wet or Dry Ice
DNA
Videos
SDS
Documents

Documents

Certificates

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.