missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ 3x DYKDDDDK Peptide

Tricosa-peptide for competitively eluting DYKDDDK-tagged fusion proteins from immobilized supports

439.00 € - 1292.00 €

Specifications

Product Line Pierce™
Product Type Peptides
Protein Form Recombinant
Format Lyophilized Powder
Shipping Condition Wet Ice
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15872684
View Documents
Thermo Scientific™
A36805
5 mg
439.00 €
5mg
In Stock
to see stock.
Add to basket
 
15882684
View Documents
Thermo Scientific™
A36806
5 x 5 mg
1292.00 €
Pack of 5
In Stock
to see stock.
Add to basket
 
Description

Description

Pierce 3X DYKDDDDK Peptide is a lyophilized peptide with the sequence Met-Asp-Tyr-Lys-Asp-His-Asp-Gly-Asp-Tyr-Lys-Asp-His-Asp-Ile-Asp-Tyr-Lys-Asp-Asp-Asp-Asp-Lys (MDYKDHDGDYKDHDIDYKDDDDK). It has a molecular weight of 2861.9 g/mol. For use, reconstitute to 1.5 mg/mL in PBS.

This tricosa-peptide can be used to competitively elute DYKDDDDK-tagged fusion proteins from immobilized anti-DYKDDDDK affinity supports, especially when a DYKDDDDK-tagged protein is sensitive to low pH. At a 1.5 mg/mL concentration, complete release of tagged protein can be achieved in 5–10 minutes at ambient temperature.

Note: Pierce 3X DYKDDDDK Peptide may interfere with protein determination assays and absorbance at 280 nm. However, the peptide can be removed from the eluate by desalting (e.g., with Zeba Spin Desalting Columns, Cat. No. 89891).

Specifications

Specifications

Pierce™
Recombinant
Wet Ice
Lyophilized Powder
Peptides
Lyophilized Powder
Protein Purification and Isolation
Affinity Resin
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Pierce™ 3x DYKDDDDK Peptide

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.