missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Ionic Detergent Compatibility Reagent for Pierce™ 660nm Protein Assay Reagent

Red-to-green (A660nm), ready-to-use, detergent- and reducing agent-compatible assay reagent to measure total protein concentration vs. protein standard.

Brand:  Thermo Scientific™ 22663

130.15 EUR valid until 2025-05-31
Use promo code "25977" to get your promotional price.


Product Code. 10570044

  • 139.00 € / 5g

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

Store between 2 to 25°C
Ionic Detergent Compatibility Reagent
Pierce™
Not Target-Specific
Spectrophotometer, Microplate Reader
Pierce Ionic Detergent Compatibility Reagent
Colorimetric
Protein Quantitation Assay
Solution-based Detection, Absorbance
5 x 1 g
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZL402506 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number22663
Lot NumberA0464711 Certificate TypeCertificate of Analysis Date25/01/2025 Catalog Number22663
Lot NumberZI397061 Certificate TypeCertificate of Analysis Date22/10/2024 Catalog Number22663
Lot NumberZH388713 Certificate TypeCertificate of Analysis Date13/09/2024 Catalog Number22663
Lot NumberZG388712 Certificate TypeCertificate of Analysis Date13/07/2024 Catalog Number22663
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Ionic Detergent Compatibility Reagent for Pierce™ 660nm Protein Assay Reagent > Compatibility Reagent; 5 x 1g

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.