missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ IPTG

Detect beta-galactosidase expression from vectors, plasmids, or DNA fragments with this chromogenic substrate.

Brand:  Thermo Scientific™ 34060

90.73 EUR valid until 2025-05-31
Use promo code "25977" to get your promotional price.


Product Code. 10658264

  • 97.50 € / 1g

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ Pierce beta-Galactosidase Precipitating Substrates is a chromogenic substrate for beta-galactosidase that is used to detect expression of vectors, plasmids, or DNA fragments that contain the reporter gene.

X-Gal (5-bromo-4-chloro-3-indolyl-β-D-galactopyranoside) is a chromogenic substrate forβ-Galactosidase. It yields a blue precipitate, is soluble in dimethylformamide, and can be used to detect vectors, plasmids or DNA fragments that contain the β-Galactosidase gene.

Highlights:

  • See item-specific features

X-Gal (5-Bromo-4-chloro-3-indolyl-β-D-galactopyranoside)

  • Chromogenic substrate for β-Galactosidase
  • Yields a blue precipitate
  • Soluble in DMF
  • Can be used to detect vectors, plasmids, or DNA fragments that contain the β-Galactosidase gene

IPTG (Isopropyl-β-D-thiogalactopyranoside)

  • Maximizes the expression of genes in expression vectors
  • When screening a lambda bacteriophage library with an antibody, the probability of success can be increased by preventing the expression of the fusion protein until the plaques are well established
  • By placing a nitrocellulose filter containing the inducer IPTG onto the plate and continuing growth at 37°C, the expression of the potential β-galactosidase-cDNA fusion protein can be induced after three to four hours of plaque growth; the nitrocellulose filter is then screened with antibodies
  • IPTG is also used in combination with X-Gal to detect active β-galactosidase
  • Practical in applications using Bacteriophage M13 and Igt11 vectors
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store at or below -20°C.
Powder
1 g
IPTG (Isopropyl β-D-thiogalactopyranoside)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZE383097 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number34060
Lot NumberYJ383096 Certificate TypeCertificate of Analysis Date21/10/2023 Catalog Number34060
Lot NumberSPECS Certificate TypeCertificate of Analysis Date06/10/2023 Catalog Number34060
Lot NumberYE3798612 Certificate TypeCertificate of Analysis Date06/10/2023 Catalog Number34060
Lot NumberYA3651741 Certificate TypeCertificate of Analysis Date13/01/2023 Catalog Number34060
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ IPTG > 1g

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.