Learn More
Thermo Scientific™ BMOE (bismaleimidoethane)
Crosslink sulfhydryls to sulfhydryls (cysteine residues in proteins or peptides) with this short crosslinker composed of maleimide groups and a 7-atom (8.0A) spacer arm.
Brand: Thermo Scientific™ 22323
161.50 EUR valid until 2025-05-31
Use promo code "25977" to get your promotional price.
Description
Thermo Scientific™ Pierce BMOE is a short-arm, maleimide crosslinker for covalent, irreversible conjugation between sulfhydryl groups (e.g., protein or peptide cysteines).
Highlights:
- Reactive groups: maleimide (homobifunctional)
- Reactive toward: sulfhydryl groups

Specifications
Sulfhydryl-Sulfhydryl | |
BMOE | |
No | |
Upon receipt store at 4°C. | |
Ambient | |
Chemical Labeling | |
Maleimide | |
Solid | |
DMF, DMSO | |
No |
No | |
220.18 | |
8.0 Å | |
Yes | |
Pierce™ | |
Homobifunctional | |
Short (<10 Å) | |
50 mg | |
Standard | |
Crosslinker |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.