missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ DNase I Solution (2500 U/mL)

Improve protein extraction efficiency with this glycosylated polypeptide that cleaves single- and double-stranded DNA in cell lysates.

Brand:  Thermo Scientific™ 90083

129.20 EUR valid until 2025-05-31
Use promo code "25977" to get your promotional price.


Product Code. 10700595

  • 140.00 € / 0.50mL

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

Remove unwanted DNA from cell lysates to improve protein extraction efficiency with Thermo Scientific™ DNase I.

Deoxyribonuclease I (DNase I) is a single, glycosylated polypeptide that degrades unwanted single- and double-stranded DNA. The enzyme works by cleaving DNA into 5' phosphodinucleotide and small oligonucleotide fragments. DNase I is commonly added to cell lysis reagents to remove the viscosity caused by the DNA content in bacterial cell lysates or to remove the DNA templates from RNAs produced by in vitro transcription. Two grades of DNAse are offered, one sufficient for protein work and one that is useful for any application requiring the digestion of DNA in which it is crucial to avoid damage to RNA.

Highlights:

  • Degrades and removes unwanted DNA from samples
  • Cleaves both single-stranded and double-stranded DNA
  • Compatible with Thermo Scientific Pierce Cell Lysis Reagents
  • Reduces viscosity of bacterial lysates (protein extracts) to facilitate pipetting
  • Supplied as 50% glycerol stocks in 10mM Tris-HCl pH 7.5, 10mM CaCl2, 10mM MgCl2
  • Available in two convenient formats: RNase-tested to protect RNA (Part No. 89835) and convenient protein extraction grade (Part No. 90083)

Recommended for:

Elimination of viscosity caused by DNA in protein cell lysates; Removal of DNA templates from RNAs produced by in vitro transcription

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Cell Lysis Enzyme
Liquid
0.5 mL
Store at -20°C
DNase
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZK402870 Certificate TypeCertificate of Analysis Date16/12/2024 Catalog Number90083
Lot NumberZK402871 Certificate TypeCertificate of Analysis Date16/12/2024 Catalog Number90083
Lot NumberZJ402869 Certificate TypeCertificate of Analysis Date26/10/2024 Catalog Number90083
Lot NumberZI394862 Certificate TypeCertificate of Analysis Date15/10/2024 Catalog Number90083
Lot NumberZG394863 Certificate TypeCertificate of Analysis Date13/08/2024 Catalog Number90083
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ DNase I Solution (2500 U/mL) > Enzyme solution; no glycerol; protein extraction grade; 0.5mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.