missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Pierce™ DSG, No-Weigh™ Format
152.00 € - 162.00 €
Specifications
Chemical Reactivity | Amine-Amine |
---|---|
Cleavable | No |
Molecular Weight (g/mol) | 326.26 |
PEGylated | No |
Spacer Arm Length | 7.7 Å |
Description
Thermo Scientific™ Pierce DSG is disuccinimidyl glutarate, a simple homobifunctional crosslinker based on the amine-reactive N-hydroxysuccinimide (NHS) ester group.
Highlights:
- Can increase crosslinking efficiency compared to DSS in some applications
- Reactive groups: NHS ester (homobifunctional)
- Reactive toward: amino groups
Specifications
Amine-Amine | |
326.26 | |
7.7 Å | |
Chemical Labeling | |
NHS Ester | |
Powder | |
Standard, Single-use | |
Crosslinker |
No | |
No | |
Yes | |
Homobifunctional | |
Short (<10 Å) | |
DMF, DMSO | |
No |
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title