missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Negative Ion Calibration Solution

Calibrate LTQ mass spectrometer instruments using a mixture of highly purified ionizable molecules designed for negative mode calibration.

Brand:  Thermo Scientific™ 88324

283.10 EUR valid until 2025-05-31
Use promo code "25977" to get your promotional price.



Product Code. 11360360

  • 301.00 € / 10mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ Pierce ESI Negative Ion Calibration Solution is a mixture of highly purified ionizable molecules designed for negative mode calibration of Thermo Scientific™ LTQ series mass spectrometer instruments.

The Pierce ESI Negative Ion Calibration Solution is a ready-to-use liquid formulation ideal for quickly performing the routine calibration required to maintain the robust performance of Thermo Scientific Mass Spectrometers. The ESI Negative Ion Calibration Solution is manufactured at an ISO 9001 facility and each lot is quality controlled with strict specifications. The stable solution is provided in a leak-proof, high-purity PTFE bottle.

Highlights:

  • Strong peaks – mixture of SDS, sodium taurocholate and Ultramark 1621 in a acetonitrile/methanol/acetic solution

  • Ready-to-use – load the mass reference standard into a syringe and inject into the instrument

  • High-purity – mass spectrometry grade reagents in a non-leachable container

  • Stable – store at room temperature for more than one year

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Mass Spectrometer
Pierce™
Peptides
Mass Spectrometry
10 mL
Calibration Solution
Store between 2°C and 8°C.
Peptides
Calibration
Liquid
Product Suggestions

Product Suggestions

Videos
Safety and Handling
Show More
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberAB407989 Certificate TypeCertificate of Analysis Date15/04/2025 Catalog Number
Lot NumberAC407406 Certificate TypeCertificate of Analysis Date28/03/2025 Catalog Number
Lot NumberAA407407 Certificate TypeCertificate of Analysis Date01/02/2025 Catalog Number
Lot NumberZJ407405 Certificate TypeCertificate of Analysis Date16/12/2024 Catalog Number88324
Lot NumberZK403449 Certificate TypeCertificate of Analysis Date16/12/2024 Catalog Number88324
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Pierce™ Negative Ion Calibration Solution > ESI Negative Ion Calibration Solution

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.