missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Horseradish Peroxidase

Purified horseradish peroxidase enzyme for use in activity assays and conjugation to antibodies for ELISA, Western blot and immunohistochemistry applications.

Brand:  Thermo Scientific™ 31491

361.95 EUR valid until 2025-04-30
Use promo code "24155" to get your promotional price.


 View more versions of this product

Product Code. 11869150

  • 391.00 € / 100mg

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

100 mg
Powder
Pierce™
Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, but do not store above -20°C.
Western Blotting, ELISA, Immunohistochemistry
HRP (Horseradish Peroxidase)
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZK403568 Certificate TypeCertificate of Analysis Date16/12/2024 Catalog Number31491
Lot NumberZI402847 Certificate TypeCertificate of Analysis Date15/10/2024 Catalog Number31491
Lot NumberZG391181 Certificate TypeCertificate of Analysis Date07/08/2024 Catalog Number31491
Lot NumberZD387385 Certificate TypeCertificate of Analysis Date09/05/2024 Catalog Number31491
Lot NumberXJ3495454 Certificate TypeCertificate of Analysis Date02/11/2022 Catalog Number31491
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Pierce™ Horseradish Peroxidase > Quantity: 100 mg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.