missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Mouse IgG1 Fab and F(ab')2 Micro Preparation Kit

Efficiently digest mouse IgG1 antibodies into Fab or F(ab')2 fragments with immobilized ficin protease.

Brand:  Thermo Scientific™ 44680

Product Code. 10721965

  • 716.00 € / 1 set

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ Pierce Mouse IgG1; Fab and F(ab')2 Preparation Kits use immobilized ficin protease to efficiently digest mouse IgG1 into Fab or F(ab')2 fragments, depending on the chosen reaction conditions.

The kits use ficin protease to enzymatically digest the Fc portion of whole IgG to yield Fab or F(ab')2 fragments. Because the ficin protease is supplied in immobilized form as beaded agarose resin, the digestion reaction is easily stopped by removing the resin from the IgG solution; the result is digest products that are enzyme-free. An optimized procedure and high-performance desalting and Protein A spin columns make these kits easy to use.

Highlights:

  • Enzyme-free digestion products – immobilized ficin (beaded agarose resin) provides for control of the digestion reaction and complete removal of resulting antibody fragments from the protease

  • Optimized for mouse IgG1 – the kit procedure is specifically optimized for monoclonal mouse IgG1 antibodies, providing the best possible yield for this antibody subclass

  • High capacity – kit provides for digestion and purification from as much as 4mg of IgG at a time. Use the standard size kit for fragmenting 0.25 to 4mg IgG, and the micro kit for 25 to 250μg IgG

  • Complete and easy to use – kits include all reagents, spin columns and accessories needed to prepare and purify antibody fragments

Includes:

Ficin agarose resin, cysteine, digestion buffer, Protein A columns with binding and elution buffers, spin desalting columns and accessories

TRUSTED_SUSTAINABILITY
Specifications

Specifications

10 Antibody Samples
Pierce™
Sufficient For: 10 antibody samples, each containing 25 to 250 μg IgG
• Pierce Ficin Agarose, 0.8 mL
• Cysteine-HCl, 1 g
• Mouse IgG1 Digestion Buffer, 120 mL
• NAb Protein A Plus Spin Columns, 0.2 mL, 2 columns
• Protein A Binding Buffer, 50 mL
• IgG Elution Buffer, 50 mL
• Zeba Spin Desalting Columns, 7K MWCO, 0.5 mL, 10 columns
• Spin Columns, 0.8 mL, 10 columns
• Microcentrifuge tubes, 2 mL, 30 tubes

Store at 4–8°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZI401449 Certificate TypeCertificate of Analysis Date15/10/2024 Catalog Number44680
Lot NumberYE371320 Certificate TypeCertificate of Analysis Date18/07/2023 Catalog Number44680
Lot NumberYE371215 Certificate TypeCertificate of Analysis Date20/05/2023 Catalog Number44680
Lot NumberYA361913 Certificate TypeCertificate of Analysis Date11/01/2023 Catalog Number44680
Lot NumberWJ335508 Certificate TypeCertificate of Analysis Date02/11/2021 Catalog Number44680
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Pierce™ Mouse IgG1 Fab and F(ab')2 Micro Preparation Kit > Pierce™ Mouse IgG1 Fab and F(ab')2 Micro Preparation Kit

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.