missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Peptide Retention Standard

A mixture of five specific deca-peptides (acetylated with c-terminal amides) to run reference elution profiles for reverse-phase HPLC chromatography.

Brand:  Thermo Scientific™ 31700

392.35 EUR valid until 2025-05-31
Use promo code "25977" to get your promotional price.


Product Code. 10361394

  • 418.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ Pierce Peptide Retention Standard is a mixture of five c-terminal amide decapeptides that provide reference elution profiles in reverse-phase HPLC.

Retention times are predicted by totaling the values that represent the contribution in minutes of each amino acid residue and the peptide terminal groups. Retention time is dependent upon the molecular weight of the peptide. The effect on retention is relatively unimportant with a small peptide, but it increases with the size of the molecule. The accuracy of predicting peptide retention time significantly decreases beyond 20 residues. To ensure accuracy, a peptide standard is used to correct for instrument variation, column aging, n-alkyl chain length variation and ligand density.

Highlights:

  • Save time in peptide purification
  • Simplify identification of specific peptides in a complex mixture
  • Increase the efficiency of predicting peptide elution profiles Determine the relative order of peptide elution of a complex mixture Predict the HPLC retention time for peptides of known amino acid composition on reverse phase HPLC columns Monitor column performance: efficiency, selectivity and resolution during column aging Compare reverse-phase columns from different manufacturers Evaluate reverse-phase supports of varying n-alkyl chain lengths and ligand densities
TRUSTED_SUSTAINABILITY
Specifications

Specifications

HPLC
Upon receipt store at -20°C. Product is shipped at ambient temperature.
HPLC
1 Vial
Peptide Retention Standard
Peptides
Standardization
Solid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZJ4448241 Certificate TypeCertificate of Analysis Date01/11/2024 Catalog Number31700,TS-31700
Lot NumberZJ4437071 Certificate TypeCertificate of Analysis Date15/10/2024 Catalog Number31700,TS-31700
Lot NumberZH4160442 Certificate TypeCertificate of Analysis Date10/08/2024 Catalog Number31700,TS-31700
Lot NumberSPECS Certificate TypeCertificate of Analysis Date11/07/2024 Catalog Number31700,TS-31700
Lot NumberZB4231021 Certificate TypeCertificate of Analysis Date08/03/2024 Catalog Number31700,TS-31700
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Peptide Retention Standard > Standard, S1-S5

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.