Learn More
Thermo Scientific™ Pierce™ Phosphatase Inhibitor Mini Tablets
Thermo Scientific™ Pierce Phosphatase Inhibitor Mini Tablets provide complete cell lysate and biological sample protection from dephosphorylation with a broad spectrum phosphatase inhibitor formulation.
Brand: Thermo Scientific™ A32957
163.30 EUR valid until 2024-12-31
Use promo code "21651" to get your promotional price.
Alert:
For Research Use Only. Not for use in diagnostic procedures.
Description
Pierce Phosphatase Inhibitor Mini Tablets protect phosphoproteins from serine/threonine phosphatases and tyrosine phosphatases. Each Phosphatase Inhibitor Tablet contains sodium fluoride, sodium orthovanadate, sodium pyrophosphate and beta-glycerophosphate for broad-spectrum inhibition of phosphatase activity. Tablets are stable at 4°C for up to 12 months, and each tablet is sufficient for 10mL of solution. The Pierce Phosphatase Inhibitor Tablets are suitable for use with any cell or tissue extract to preserve protein phosphorylation for downstream applications.
Highlights:- Economical&mdsashmore cost effective than other commercially available formulations
- Improved formulation&mdsashtablets dissolve quickly into a clear solution
- Complete&mdsashthe tablets inhibit acid, alkaline, and protein phosphatases
- Compatible&mdsashuse with homebrew or commercially available cell lysis reagents and Thermo Scientific Protein Assays
Specifications
Protease Inhibitor Cocktail | |
Wet Ice | |
Pierce™ | |
Phosphatase Inhibitor | |
Sodium Fluoride, Sodium Orthovanadate, Sodium Pyrophosphate, Beta-Glycerophosphate |
1 Vial, Store at 2°C to 8°C | |
Solid | |
10 mL Solution | |
20 Tablets |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.