Learn More
Thermo Scientific™ Pierce™ Phosphatase Inhibitor Mini Tablets
Thermo Scientific™ Pierce Phosphatase Inhibitor Mini Tablets provide complete cell lysate and biological sample protection from dephosphorylation with a broad spectrum phosphatase inhibitor formulation.
26.60 € - 241.00 €
Alert:
For Research Use Only. Not for use in diagnostic procedures.
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15691759
View Documents
Thermo Scientific™
A32957
20 Tablets
241.00 €
Pack of 20
In Stock
to see stock.
Edge
Add to basket
16306532
View Documents
Thermo Scientific™
A32958
2 Tablets
26.60 €
Each
Estimated Shipment: 27-01-2025
to see stock.
Add to basket
Description
Description
Pierce Phosphatase Inhibitor Mini Tablets protect phosphoproteins from serine/threonine phosphatases and tyrosine phosphatases. Each Phosphatase Inhibitor Tablet contains sodium fluoride, sodium orthovanadate, sodium pyrophosphate and beta-glycerophosphate for broad-spectrum inhibition of phosphatase activity. Tablets are stable at 4°C for up to 12 months, and each tablet is sufficient for 10mL of solution. The Pierce Phosphatase Inhibitor Tablets are suitable for use with any cell or tissue extract to preserve protein phosphorylation for downstream applications.
Highlights:
  • Economical—more cost effective than other commercially available formulations
  • Improved formulation—tablets dissolve quickly into a clear solution
  • Complete—the tablets inhibit acid, alkaline, and protein phosphatases
  • Compatible—use with homebrew or commercially available cell lysis reagents and Thermo Scientific Protein Assays