Learn More
Thermo Scientific™ Pierce™ Protease Inhibitor Mini Tablets
Thermo Scientific™ Pierce Protease Inhibitor Mini Tablets are broad-spectrum protease inhibitors that are highly effective at preventing proteolytic degradation for cell lysis and protein extraction experiments
Brand:  Thermo Scientific™ A32954
 View more versions of this product
Product Code. 16386522
  • 26.53 € / Each
Estimated Shipment: 24-01-2025
to see stock.
Explore more special offers
Add to basket
Alert:
For Research Use Only. Not for use in diagnostic procedures.
Description
Description
Pierce Protease Inhibitor Tablets protect proteins from proteolytic degradation through inhibition of the serine-proteases, cysteine-proteases, aspartic acid-proteases and aminopeptidases that are typically present in cellular lysate samples. The tablets contain AEBSF, aprotinin, bestatin, E-64, leupeptin and pepstatin A. Stable for storage at 4°C for up to 12 months, each mini or large tablet tablet is sufficient for 10mL or 50mL of solution, respectively, and is suitable for reagent preparation prior to protein extraction from tissue and cultured cells.
Highlights:
  • Improved formulation&mdsashtablets dissolve quickly into a clear solution
  • Economical – more cost-effective than other commercially available formulations for equivalent volumes
  • Convenient&mdsashready-to-use, fully disclosed, broad-spectrum formulations
  • Compatible&mdsashuse directly with Thermo Scientific™ Pierce™ BCA Assays
TRUSTED_SUSTAINABILITY
Specifications
Specifications
EDTA
protein purification, western blotting
2 Tablets
2°C to 8°C
1 tablet is sufficient for 10 mL
Tablet