missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Protease Inhibitor Mini Tablets

Thermo Scientific™ Pierce Protease Inhibitor Mini Tablets are broad-spectrum protease inhibitors that are highly effective at preventing proteolytic degradation for cell lysis and protein extraction experiments

26.53 € - 26.62 €

Specifications

Content And Storage 2°C to 8°C
For Use With (Application) protein purification, western blotting
Sufficient For 1 tablet is sufficient for 10 mL
Quantity 2 Tablets
Format Tablet
View More Specs

Alert:

For Research Use Only. Not for use in diagnostic procedures.


Products 2
Product Code Brand Includes Price Quantity & Availability  
Product Code Brand Includes Price Quantity & Availability  
16386522
View Documents
Thermo Scientific™
A32954
EDTA
26.53 €
Each
Estimated Shipment: 23-01-2025
to see stock.
Add to basket
 
16396522
View Documents
Thermo Scientific™
A32956
EDTA Free
26.62 €
Each
Estimated Shipment: 23-01-2025
to see stock.
Add to basket
 
Description

Description

Pierce Protease Inhibitor Tablets protect proteins from proteolytic degradation through inhibition of the serine-proteases, cysteine-proteases, aspartic acid-proteases and aminopeptidases that are typically present in cellular lysate samples. The tablets contain AEBSF, aprotinin, bestatin, E-64, leupeptin and pepstatin A. Stable for storage at 4°C for up to 12 months, each mini or large tablet tablet is sufficient for 10mL or 50mL of solution, respectively, and is suitable for reagent preparation prior to protein extraction from tissue and cultured cells.

Highlights:
  • Improved formulation—tablets dissolve quickly into a clear solution
  • Economical – more cost-effective than other commercially available formulations for equivalent volumes
  • Convenient—ready-to-use, fully disclosed, broad-spectrum formulations
  • Compatible—use directly with Thermo Scientific™ Pierce™ BCA Assays
Specifications

Specifications

2°C to 8°C
1 tablet is sufficient for 10 mL
Tablet
protein purification, western blotting
2 Tablets
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.