missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Protein A Magnetic Beads

Perform antibody purification and immunoprecipitation methods using manual or robotic magnetic separators with these high-capacity and high-throughput affinity particles.

Brand:  Thermo Scientific™ 88845

 View more versions of this product

Product Code. 13494219

  • 250.00 € / 1mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific Pierce Protein A Magnetic Beads are high-capacity and high-throughput affinity particles for antibody purification and immunoprecipitation methods using manual or robotic magnetic separators.

Features of Protein A Magnetic Beads:

  • High efficiency – equivalent or higher yield of IP target antigens than magnetic beads from other suppliers
  • Low non-specific binding – stable, pre-blocked beads provide highly purified product (e.g., antigen eluted in IP with antibody is devoid of contaminating protein from complex cell lysate)
  • Consistent – magnetic beads eliminate resin loss and provide for more efficient separation than traditional IP methods that use only centrifugation
  • Versatile – beads are compatible with manual and automated workflows (e.g., Thermo Scientific KingFisher Instruments)

Recombinant protein A, covalently attached to a blocked magnetic bead surface, can bind to antibodies from many different species, enabling purification of antibodies from crude extracts. Immunoprecipitation assays performed with protein A coated beads result in high yield of target antigen with very low background. Using our crosslinker chemistry, you can immobilize an antibody onto the magnetic particle and prevent IgG contamination in IP or Co-IP experiments. The Pierce Protein A Magnetic Beads can be used manually with a magnetic stand, as well as with automated platforms, such as the Thermo Scientific KingFisher Instruments, for high-throughput workflows.

Applications:

  • IP and Co-IP experiments
  • Antibody purification

Thermo Scientific Pierce Protein A Magnetic Beads are used for purifying antibody from serum, cell culture supernatant or ascites, as well as for IP/Co-IP of antigens from cell or tissue extracts. Protein A can bind to antibodies from many different species, including mouse, human, rabbit, pig, dog, and cat. The protocol for the Pierce Protein A beads has been optimized to allow for high recovery and high purity of the isolated antibody or antigen. Antibody or antigen/antibody complex (IP) is first captured on the magnetic beads. The beads are washed and then the target is eluted with low pH elution buffer. IP performance is equivalent to or better than Protein A magnetic beads from other suppliers.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

1 mL
Upon receipt store at 4°C. Product shipped with an ice pack.
Liquid
1 mL
Antibody
Protein A
Slurry: 10mg/mL, 1% solids
1 μm
Pierce™
Recombinant
Magnetic Affinity Bead
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZL398153 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number88845
Lot NumberZH398791 Certificate TypeCertificate of Analysis Date17/08/2024 Catalog Number88845
Lot NumberZD387758 Certificate TypeCertificate of Analysis Date17/04/2024 Catalog Number88845
Lot NumberYJ379129 Certificate TypeCertificate of Analysis Date20/10/2023 Catalog Number88845
Lot NumberYF379128 Certificate TypeCertificate of Analysis Date01/07/2023 Catalog Number88845
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Pierce™ Protein A Magnetic Beads > 1 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.