missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Recombinant Protein A/G

Detect antibodies, especially IgG isotypes, from many different species hosts with this purified, alkaline phosphatase- or peroxidase-conjugated recombinant Protein A/G.

Brand:  Thermo Scientific™ 77678

 View more versions of this product

Product Code. 15215693

  • 12380.00 € / 500mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Purified and conjugated forms of Thermo Scientific™ Pierce Recombinant Protein A/G enable probing and detection of antibodies, especially IgG isotypes, from many different species hosts in Western blotting, ELISA, IHC and other immunoassay protocols.

The extended Fc-binding properties of Protein A/G make it a popular tool in the investigation and purification of immunoglobulins. Protein A/G binds to all human IgG subclasses, IgA, IgE, IgM and to a lesser extent IgD; however, it does not bind mouse IgA, IgM or murine serum albumin. Protein A/G is an excellent tool for purification and detection of mouse monoclonal antibodies from IgG subclasses without interference from these other serum proteins. Individual subclasses of mouse monoclonals are most likely to have stronger affinity to this chimeric protein than to either Protein A or Protein G.

Highlights:

  • Contains four Fc-binding domains from Protein A and two from Protein G
  • Provides binding for all antibody species and subclasses recognized by either Protein A or Protein G
TRUSTED_SUSTAINABILITY
Specifications

Specifications

500 mg
Powder
Antibodies
Pierce™
Protein A
Upon receipt store at -20°C. Product is shipped at ambient temperature.
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZJ403843 Certificate TypeCertificate of Analysis Date19/10/2024 Catalog Number77678
Lot NumberZH402434 Certificate TypeCertificate of Analysis Date13/08/2024 Catalog Number77678
Lot NumberZF401681 Certificate TypeCertificate of Analysis Date02/07/2024 Catalog Number77678
Lot NumberZF398831 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog Number77678
Lot NumberZC396743 Certificate TypeCertificate of Analysis Date04/04/2024 Catalog Number77678
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Pierce™ Recombinant Protein A/G > 500mg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.