missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Recombinant Protein G

Detect mouse and human antibodies, especially IgG isotypes, in immunoassays with this purified, biotinylated or peroxidase-conjugated recombinant Protein G.

Brand:  Thermo Scientific™ 77675

 View more versions of this product

Product Code. 15285683

  • 1070.00 € / 50mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Purified and conjugated forms of Thermo Scientific™ Pierce Recombinant Protein G enable probing and detection of mouse and human antibodies, especially IgG isotypes, in Western blotting, ELISA, IHC and other immunoassay protocols.

Protein G is a bacterial cell wall protein isolated from group G Streptococci. DNA sequencing of native Protein G identifies two IgG-binding domains and sites for albumin and cell surface binding. The albumin and cell surface binding domains have been eliminated from Recombinant Protein G to reduce nonspecific binding and, therefore, can be used to separate IgG from crude samples. Optimal binding occurs at pH 5, although binding is also effective at pH 7.0 to 7.2.

Highlights:

  • Contains two Fc-binding domains per protein
  • Better than Protein A for mouse (incl. IgG1), human (incl. IgG3) and rat, goat and cow antibodies
  • Poorer than Protein A for guinea pig, pig, dog and cat
  • Does not bind human IgM, IgD or IgA
TRUSTED_SUSTAINABILITY
Specifications

Specifications

50 mg
Powder
Antibodies
Pierce™
Protein G
Upon receipt store at -20°C. Product is shipped at ambient temperature.
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZI400999 Certificate TypeCertificate of Analysis Date15/10/2024 Catalog Number77675
Lot NumberZC388604 Certificate TypeCertificate of Analysis Date19/03/2024 Catalog Number77675
Lot NumberYD365873 Certificate TypeCertificate of Analysis Date04/05/2023 Catalog Number77675
Lot NumberXI364180 Certificate TypeCertificate of Analysis Date08/10/2022 Catalog Number77675
Lot NumberXC339969 Certificate TypeCertificate of Analysis Date17/05/2022 Catalog Number77675
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Pierce™ Recombinant Protein G > 50mg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.