missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences WHEATON™ COMPLETEPAK MODPAK Amber Kit

Range of crimp top vials, rubber stoppers and seals that are sterilized ready-to-use and are off- the-shelf, and can be customized for any application reducing your supply chain due to our one source solution capability.

Brand:  DWK Life Sciences W991012A-CPMP

 View more versions of this product

Product Code. 17962671

  • 7255.00 € / 1 set
Estimated Shipment: 12-06-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Six inventory matched kits conveniently packed into a single shipper case.
  • Sterilized, off-the-shelf, ready-to-use (RtU) primary packaging components
  • Focuses your effort on core competencies and reduces lead time
  • Streamlines process and frees up valuable clean room space USP /EP 2.9.19
  • Particulate Matter in Injections, USP /EP 2.6.1,
  • The Bacterial Endotoxins Test and USP / EP 2.6.14
  • The Sterility Test, USP /EP 3.2.1
  • Glass Containers for Pharmaceutical Use
  • USP /EP 3.2.9 Elastomeric Closures used in Parenteral Preparations
  • Complete manufacturing & certificate of analysis traceability
  • Ready-to-Use, Just-in-Time
  • Minimum 180 day shelf life from time of shipment
  • Container Closure Integrity (CCI) validation
  • Available in amber for photosensitive contents
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Blue
24 mm
Single-use
Compounding pharmaceuticals, Aseptic filling of injectable drug products, Drug discovery, Drug research, Freeze drying / lyophilized drug products, Large and small molecule drug packaging
Vial, Stopper, and Seal
Bromobutyl
Ultra Pure Serum Stopper
104/Pk.
Serum Vial
Amber
Sterile
10 mL
No
CompletePAK ModPAK
Autoclavable
50 mm
USP <788>/EP 2.9.19 Particulate Matter in Injections, USP <85>/EP 2.6.1, The Bacterial Endotoxins Test and USP <71>/ EP 2.6.14 The Sterility Test, USP <660>/EP 3.2.1 Glass Containers for Pharmaceutical Use, and USP <381>/EP 3.2.9 Elastomeric Closures used in Parenteral Preparations
20 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
DWK Life Sciences WHEATON™ COMPLETEPAK MODPAK Amber Kit > Vial Capacity: 10 mL,Stopper: 20 mm Ultra Pure Serum, Seal:Blue Seal

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.